missing translation for 'onlineSavingsMsg'
Learn More

Saccharin Sodium (Powder/USP/FCC), Fisher Chemical™

Catalog No. p-191320
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S394500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S394500 Supplier Thermo Fisher Scientific Supplier No. FLS394 500
Only null left

Chemical Identifiers

CAS 128-44-9
Molecular Formula C7H9NNaO5S
Molecular Weight (g/mol) 242.201
MDL Number MFCD00013092
InChI Key UQKRDMFNUSMUCB-UHFFFAOYSA-N
Synonym saccharin sodium dihydrate, saccharin sodium, sodium saccharin, saccharin soluble, saccharin sodium salt hydrate, 1,2-benzisothiazol-3 2h-one, 1,1-dioxide, sodium salt, dihydrate, o-sulfobenzimide sodium salt, 2-sulfobenzoic acid imide sodium salt, saccharin sodium salt dihydrate, c7h4nnao3s.2h2o
PubChem CID 46942257
IUPAC Name 1,1-dioxo-1,2-benzothiazol-3-one;sodium;dihydrate
SMILES C1=CC=C2C(=C1)C(=O)NS2(=O)=O.O.O.[Na]

Specifications

Alkalinity Pass Test (FCC)
Acidity Pass Test (USP)
Color White
Packaging Poly Bottle
Quantity 500 g
Residual Solvents Meets Requirements (USP)
Formula Weight 205.17
Percent Purity 98.0 to 101.0% (USP)
Grade FCC/USP
Physical Form Solid
Chemical Name or Material Saccharin Sodium
Show More Show Less