missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ S1P2/EDG-5/S1PR2 Antibody Blocking Peptide
Specifications
Specifications
| Gene ID (Entrez) | 9294 |
| Product Line | Novus |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| For Use With (Application) | This peptide is useful as a blocking peptide for NBP2-26691. For further blocking peptide related protocol, click here . |
| Gene Alias | AGR16, EDG5, EDG-5, endothelial differentiation G-protein coupled receptor 5, endothelial differentiation, sphingolipid G-protein-coupled receptor, 5, Gpcr13, H218, LPB2, S1P receptor 2, S1P receptor Edg-5, S1P receptor EDG5, S1P2, sphingosine 1-phosphate receptor 2, sphingosine 1-phosphate receptor Edg-5, sphingosine-1-phosphate receptor 2 |
| Gene Symbol | S1PR2 |
| Quantity | 0.1 mg |
| Research Category | GPCR |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |