missing translation for 'onlineSavingsMsg'
Learn More

(S)-Trolox methyl ether, ≥98.0% (sum of enantiomers, HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013005
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013005 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013005 Supplier Merck Emd Millipore Supplier No. 9351050MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00171453
Synonym (S)-6-Methoxy-2,5,7,8-tetramethylchromane-2-carboxylic acid

Specifications

Melting Point 143°C to 145°C
CAS 135806-59-6
Packaging Bottomless Glass Bottle with Inserted Fused Cone
Linear Formula C15H20O4
Quantity 50 mg
Formula Weight 264.32
Percent Purity ≥98.0% (sum of enantiomers, HPLC); ≥98.0%
Grade For chiral derivatization