missing translation for 'onlineSavingsMsg'
Learn More

(-)-S-Trityl-D-cysteine, 98%

CAS: 25840-82-8 | C22H21NO2S | 363.48 g/mol

$102.92 - $391.90

Chemical Identifiers

CAS 25840-82-8
Molecular Formula C22H21NO2S
Molecular Weight (g/mol) 363.48
MDL Number MFCD00236948
InChI Key DLMYFMLKORXJPO-UHFFFAOYNA-N
Synonym h-d-cys trt-oh, s-trityl-d-cysteine, h-cys trt-oh, s-2-amino-3-tritylthio propanoic acid, 2s-2-amino-3-triphenylmethyl sulfanyl propanoic acid, d-cysteine, s-triphenylmethyl, ambotzhaa6120, h-cys trt-2-chlorotrityl resin, --s-trityl-d-cysteine, 2s-2-amino-3-tritylsulfanylpropanoic acid
PubChem CID 7271796
IUPAC Name 2-amino-3-[(triphenylmethyl)sulfanyl]propanoic acid
SMILES NC(CSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6302203
View Documents
Thermo Scientific Chemicals
H6302203
1 g N/A
Only null left
 
AAH6302206
View Documents
Thermo Scientific Chemicals
H6302206
5 g N/A
Only null left
 
Description

Description

(-)-S-Trityl-D-cysteine is used in medicine as intermediate. It inhibits the human mitotic kinesin Eg5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(-)-S-Trityl-D-cysteine is used in medicine as intermediate. It inhibits the human mitotic kinesin Eg5.

Solubility
Sparingly soluble in water 0.022 g/L 25°C.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, heat.
Specifications

Chemical Identifiers

25840-82-8
363.48
DLMYFMLKORXJPO-UHFFFAOYNA-N
7271796
NC(CSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O
C22H21NO2S
MFCD00236948
h-d-cys trt-oh, s-trityl-d-cysteine, h-cys trt-oh, s-2-amino-3-tritylthio propanoic acid, 2s-2-amino-3-triphenylmethyl sulfanyl propanoic acid, d-cysteine, s-triphenylmethyl, ambotzhaa6120, h-cys trt-2-chlorotrityl resin, --s-trityl-d-cysteine, 2s-2-amino-3-tritylsulfanylpropanoic acid
2-amino-3-[(triphenylmethyl)sulfanyl]propanoic acid

Specifications

25840-82-8
MFCD00236948
Sparingly soluble in water 0.022g/L 25°C.
NC(CSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O
1 g
7271796
98%
(-)-S-Trityl-D-cysteine
C22H21NO2S
h-d-cys trt-oh, s-trityl-d-cysteine, h-cys trt-oh, s-2-amino-3-tritylthio propanoic acid, 2s-2-amino-3-triphenylmethyl sulfanyl propanoic acid, d-cysteine, s-triphenylmethyl, ambotzhaa6120, h-cys trt-2-chlorotrityl resin, --s-trityl-d-cysteine, 2s-2-amino-3-tritylsulfanylpropanoic acid
DLMYFMLKORXJPO-UHFFFAOYNA-N
2-amino-3-[(triphenylmethyl)sulfanyl]propanoic acid
363.48
363.47
Solid
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only