missing translation for 'onlineSavingsMsg'
Learn More

(-)-S-Trityl-D-cysteine, 98%

Catalog No. AAH6302206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6302206 5 g
AAH6302203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6302206 Supplier Thermo Scientific Chemicals Supplier No. H6302206

CAS: 25840-82-8 | C22H21NO2S | 363.48 g/mol

(-)-S-Trityl-D-cysteine is used in medicine as intermediate. It inhibits the human mitotic kinesin Eg5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(-)-S-Trityl-D-cysteine is used in medicine as intermediate. It inhibits the human mitotic kinesin Eg5.

Solubility
Sparingly soluble in water 0.022 g/L 25°C.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, heat.

Chemical Identifiers

CAS 25840-82-8
Molecular Formula C22H21NO2S
Molecular Weight (g/mol) 363.48
MDL Number MFCD00236948
InChI Key DLMYFMLKORXJPO-UHFFFAOYNA-N
Synonym h-d-cys trt-oh, s-trityl-d-cysteine, h-cys trt-oh, s-2-amino-3-tritylthio propanoic acid, 2s-2-amino-3-triphenylmethyl sulfanyl propanoic acid, d-cysteine, s-triphenylmethyl, ambotzhaa6120, h-cys trt-2-chlorotrityl resin, --s-trityl-d-cysteine, 2s-2-amino-3-tritylsulfanylpropanoic acid
PubChem CID 7271796
IUPAC Name 2-amino-3-[(triphenylmethyl)sulfanyl]propanoic acid
SMILES NC(CSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O

Specifications

Solubility Information Sparingly soluble in water 0.022g/L 25°C.
Quantity 5 g
Formula Weight 363.47
Percent Purity 98%
Physical Form Solid
Chemical Name or Material (-)-S-Trityl-D-cysteine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only