missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Tetrahydrofuran-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. T17415G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17415G 5 g
T17411G 1 g
T174125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T17415G Supplier TCI America Supplier No. T17415G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 87392-07-2
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.116
MDL Number MFCD00272639
InChI Key UJJLJRQIPMGXEZ-BYPYZUCNSA-N
Synonym s-tetrahydrofuran-2-carboxylic acid, s---2-tetrahydrofuroic acid, s---tetrahydro-2-furoic acid, 2s-oxolane-2-carboxylic acid, s-tetrahydro-2-furoic acid, s---tetrahydrofuran-2-carboxylic acid, unii-0i2b46k2sj, tetrahydro-2-furoic acid,-, 2-furancarboxylic acid, tetrahydro-, 2s, s---2-carboxytetrahydrofuroic acid
PubChem CID 446666
ChEBI CHEBI:45960
IUPAC Name (2S)-oxolane-2-carboxylic acid
SMILES C1CC(OC1)C(=O)O

Specifications

Color Yellow
Quantity 5 g
Physical Form Clear Liquid at 20°C
UN Number 3265
Boiling Point 251°C
Formula Weight 116.12
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material (S)-(-)-Tetrahydrofuran-2-carboxylic Acid