missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Tetrahydro-3-furoic acid, 97%

Catalog No. p-7057062
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66392MD 250 mg
AAH6639203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66392MD Supplier Thermo Scientific Chemicals Supplier No. H66392MD
Only null left

CAS: 168395-26-4 | C5H8O3 | 116.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 168395-26-4
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.12
MDL Number MFCD07369984
InChI Key BOTREHHXSQGWTR-UHFFFAOYNA-N
Synonym s-tetrahydrofuran-3-carboxylic acid, 3s-oxolane-3-carboxylic acid, s-tetrahydro-3-furancarboxylic acid, s-tetrahydro-3-furoic acid, s-tetrahydro-furan-3-carboxylic acid, 3s-tetrahydrofuran-3-carboxylic acid, l-tetrahydrofuran-3-carboxylic acid, 3-furancarboxylic acid, tetrahydro-, 3s, 3s-3-oxolanecarboxylic acid, s, ?-tetrahydro-3-furancarboxylic acid
PubChem CID 40784874
IUPAC Name oxolane-3-carboxylic acid
SMILES OC(=O)C1CCOC1

Specifications

Color Yellow to Brown
Quantity 250 mg
Beilstein 1281203
Formula Weight 116.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (S)-(-)-Tetrahydro-3-furoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only