missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-tert-Butylsulfinamide 98.0+%, TCI America™

Catalog No. B29085G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29085G 5 g
B29081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29085G Supplier TCI America Supplier No. B29085G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 343338-28-3
Molecular Formula C4H11NOS
Molecular Weight (g/mol) 121.198
MDL Number MFCD05861480
InChI Key CESUXLKAADQNTB-ZETCQYMHSA-N
Synonym s---2-methyl-2-propanesulfinamide, s-2-methylpropane-2-sulfinamide, s-tert-butanesulfinamide, s---tert-butanesulfinamide, s---t-butylsulfinamide, s---tert-butylsulfinamide, s-2-methyl-2-propanesulfinamide, s-tert-butylsulfinamide, s---t-butylmethylsulfinamide, s-t-butyl sulfinamide
PubChem CID 11355477
IUPAC Name 2-methylpropane-2-sulfinamide
SMILES CC(C)(C)S(=O)N

Specifications

Melting Point 106°C
Color White-Yellow
Quantity 5 g
Formula Weight 121.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-tert-Butylsulfinamide