missing translation for 'onlineSavingsMsg'
Learn More

(S)-tert-Butylsulfinamide, 97+%, (>98.0% ee), Thermo Scientific Chemicals

Catalog No. p-3598221
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC391080050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC391080050 Supplier Thermo Scientific Chemicals Supplier No. 391080050
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 343338-28-3
Molecular Formula C4H11NOS
Molecular Weight (g/mol) 121.2
MDL Number MFCD05861480
InChI Key CESUXLKAADQNTB-ZETCQYMHSA-N
Synonym s---2-methyl-2-propanesulfinamide, s-2-methylpropane-2-sulfinamide, s-tert-butanesulfinamide, s---tert-butanesulfinamide, s---t-butylsulfinamide, s---tert-butylsulfinamide, s-2-methyl-2-propanesulfinamide, s-tert-butylsulfinamide, s---t-butylmethylsulfinamide, s-t-butyl sulfinamide
PubChem CID 11355477
IUPAC Name 2-methylpropane-2-sulfinamide
SMILES CC(C)(C)S(=O)N

Specifications

CAS Min % 97.0
CAS Max % 100.0
Melting Point 101°C
Color White
Enantiomeric Excess 98.0% min.
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CSONH2
Quantity 5 g
Merck Index 15, 3607
Formula Weight 121.2
Percent Purity 97+%
Physical Form Crystalline Powder
Chemical Name or Material (S)-tert-Butylsulfinamide
Show More Show Less

RUO – Research Use Only