missing translation for 'onlineSavingsMsg'
Learn More

S-tert-Butyl-L-cysteine hydrochloride, 98%

Catalog No. AAH5924406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5924406 5 g
AAH5924414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5924406 Supplier Thermo Scientific Chemicals Supplier No. H5924406
Only null left

CAS: 2481-09-6 | C7H16ClNO2S | 213.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2481-09-6
MDL Number MFCD00037426

Specifications

Color White
Molecular Formula C10H16Si2
Synonym 1,4-bis dimethylsilyl benzene, benzene, 1,4-bis dimethylsilyl, p-phenylenebis dimethylsilane, 1,4-phenylenebis dimethylsilane, p-bis dimethylsilyl benzene, silane, p-phenylenebis dimethyl, silane, 1,4-phenylenebis dimethyl, 1,4 bis dimethylsilyl benzene
InChI Key UHXCHUWSQRLZJS-UHFFFAOYSA-N
SMILES C[Si](=C1C=CC(=[Si](C)C)C=C1)C
IUPAC Name (4-dimethylsilylidenecyclohexa-2,5-dien-1-ylidene)-dimethylsilane
Quantity 5 g
Molecular Weight (g/mol) 192.41
PubChem CID 6328729
Formula Weight 213.73
Percent Purity 98%
Physical Form Powder
Chemical Name or Material S-tert-Butyl-L-cysteine hydrochloride
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only