missing translation for 'onlineSavingsMsg'
Learn More

(S)-Styrene oxide, 94%

Catalog No. p-4812530
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC449450050 5 g Glass bottle
AC449450250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC449450050 Supplier Thermo Scientific Chemicals Supplier No. 449450050
Only null left

CAS: 20780-54-5 | C8H8O | 120.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20780-54-5
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.15
MDL Number MFCD00064310,MFCD00066210
InChI Key AWMVMTVKBNGEAK-MRVPVSSYSA-N
Synonym s-styrene oxide, s-2-phenyloxirane, 2s-2-phenyloxirane, s-phenyloxirane, s-epoxyethyl benzene, s-+-styrene oxide, styrene oxide, s, unii-av5p894c84, ccris 4094
PubChem CID 114946
ChEBI CHEBI:51014
IUPAC Name (2S)-2-phenyloxirane
SMILES C1O[C@H]1C1=CC=CC=C1

Specifications

Melting Point -37.0°C
Density 1.0510g/mL
Boiling Point 192.0°C to 194.0°C
Flash Point 79°C
Assay Percent Range 92% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 1.051
Solubility Information Solubility in water: practically insoluble
Formula Weight 120.15
Percent Purity 94%
Chemical Name or Material (S)-Styrene oxide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause cancer.
Harmful in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause genetic defects.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only