missing translation for 'onlineSavingsMsg'
Learn More

3M™ S-Series Hood with Inner Shroud, Integrated Head Suspension

Catalog No. p-7246421
Change view
Click to view available options
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19039548 Large
19039549 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19039548 Supplier 3M™ Supplier No. 7000052786
Only null left

Economy hood for use with 3M™ PAPR and SAR systems. Available in small/medium or medium/large size.

  • Versaflo S-Series economy hood
  • Polypropylene
  • Air Regulating Valve, Cooling / Heating Valve, Low Pressure Valve
  • 20/Case
  • Applications include cleaning, assembly, mechanical, painting
  • 3M Catalog# S-403L-20, S-403S-20

Specifications

Assigned Protection Factor (APF) 1000
Certifications/Compliance NIOSH
Color White
Material Polypropylene
Product Type Hood with Inner Shroud
Size Large
For Use With (Application) Military maintenance, repair and operation, pharmaceutical, transportation, metal production and fabrication, automotive, food and beverage manufacturing
For Use With (Equipment) Powered air purifying and supplied air respirator systems
Includes Head, face, neck, and shoulder coverage, fabric suspension, inner shroud
Model S-403L-20
Quantity 20/Cs.
Suspension Type Integrated
Visor Type PETG
Show More Show Less