missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-sec-Butanol, 99%

Catalog No. AC218030010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC218030010 1 g
AC218030050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC218030010 Supplier Thermo Scientific Chemicals Supplier No. 218030010

CAS: 4221-99-2 | C4H10O | 74.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4221-99-2
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00064281
InChI Key BTANRVKWQNVYAZ-BYPYZUCNSA-N
Synonym s-+-2-butanol, s-2-butanol, 2s-butan-2-ol, s-butan-2-ol, s-+-sec-butanol, 2s-2-butanol, unii-69kxu5ndto, s---2-butanol, s-+-sec-butyl alcohol
PubChem CID 444683
ChEBI CHEBI:45475
IUPAC Name (2S)-butan-2-ol
SMILES CCC(C)O

Specifications

Melting Point -114°C
Color Colorless
CAS Min % 98.5
CAS Max % 100
Density 0.8000g/mL
Quantity 1 g
Flash Point 26°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3960 to 1.398
Assay Percent Range 98.5% min. (GC)
Specific Gravity 0.8
Linear Formula C2H5CH(OH)CH3
Viscosity 4.21 mPa.s (20°C)
Beilstein 01, 373
Merck Index 15, 1543
Boiling Point 99°C to 100°C
Solubility Information Solubility in water: in water: 125g/L (20°C)
Formula Weight 74.12
Percent Purity 99%
Chemical Name or Material (S)-(+)-sec-Butanol, 0.99
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 224-168-1
TSCA TSCA

RUO – Research Use Only