missing translation for 'onlineSavingsMsg'
Learn More

(S,S)-TaDiAS-2nd 95.0+%, TCI America™

Catalog No. D3476200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3476200MG 200 mg
D34761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3476200MG Supplier TCI America Supplier No. D3476200MG
Only null left

Chemical Identifiers

CAS 2135524-59-1
Molecular Formula C58H70B2F8N2O2
Synonym (S,S)-TaDiAS-2nd

Specifications

Color White-Yellow
Quantity 200 mg
UN Number 1759
Formula Weight 1000.82
Percent Purity ≥95.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 6,10-Dibenzyl-N,N′-dimethyl-N,N,N′,N′-tetrakis(4-methylbenzyl)-1,4-dioxaspiro[4.5]