missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-S-Methyl-S-phenylsulfoximine, 97%

CAS: 33903-50-3 | C7H9NOS | 155.215 g/mol

$167.34 - $443.10

Chemical Identifiers

CAS 33903-50-3
Molecular Formula C7H9NOS
Molecular Weight (g/mol) 155.215
MDL Number MFCD00151461
InChI Key YFYIDTVGWCYSEO-JTQLQIEISA-N
Synonym s-+-s-methyl-s-phenylsulfoximine, s-+-s-methyl-s-phenylsulfoximine ee, s-imino methyl phenyl-??-sulfanone, r---s-methyl-s-phenylsulfoximine ee, imino-methyl-oxo-phenyl-$l^ 6, s-+-s-methyl-s-phenylsulphoximine, sulfoximine,s-methyl-s-phenyl-, s s
PubChem CID 25036288
IUPAC Name imino-methyl-oxo-phenyl-$l^{6}-sulfane
SMILES CS(=N)(=O)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL08618MD
View Documents
Thermo Scientific Chemicals
L08618MD
250 mg
Each for $167.34
Only null left
 
AAL0861803
View Documents
Thermo Scientific Chemicals
L0861803
1 g
Each for $443.10
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

33903-50-3
155.215
YFYIDTVGWCYSEO-JTQLQIEISA-N
25036288
CS(=N)(=O)C1=CC=CC=C1
C7H9NOS
MFCD00151461
s-+-s-methyl-s-phenylsulfoximine, s-+-s-methyl-s-phenylsulfoximine ee, s-imino methyl phenyl-??-sulfanone, r---s-methyl-s-phenylsulfoximine ee, imino-methyl-oxo-phenyl-$l^ 6, s-+-s-methyl-s-phenylsulphoximine, sulfoximine,s-methyl-s-phenyl-, s s
imino-methyl-oxo-phenyl-$l^{6}-sulfane

Specifications

33903-50-3
C7H9NOS
250 mg
Hygroscopic
YFYIDTVGWCYSEO-JTQLQIEISA-N
CS(=N)(=O)C1=CC=CC=C1
155.215
155.22
(S)-(+)-S-Methyl-S-phenylsulfoximine
30°C to 32°C
MFCD00151461
5245633
s-+-s-methyl-s-phenylsulfoximine, s-+-s-methyl-s-phenylsulfoximine ee, s-imino methyl phenyl-??-sulfanone, r---s-methyl-s-phenylsulfoximine ee, imino-methyl-oxo-phenyl-$l^ 6, s-+-s-methyl-s-phenylsulphoximine, sulfoximine,s-methyl-s-phenyl-, s s
+36° (c=1 in Acetone)
imino-methyl-oxo-phenyl-$l^{6}-sulfane
25036288
97%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only