missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S,S)-2,6-Bis(4-phenyl-2-oxazolin-2-yl)pyridine 95.0+%, TCI America™
$255.38 - $743.76
Chemical Identifiers
| CAS | 174500-20-0 |
|---|---|
| Molecular Formula | C23H19N3O2 |
| Molecular Weight (g/mol) | 369.424 |
| MDL Number | MFCD01863586 |
| InChI Key | HLHBIMJNCKZZQO-NHCUHLMSSA-N |
| Synonym | 2,6-bis s-4-phenyl-4,5-dihydrooxazol-2-yl pyridine, 2,6-bis 4s-phenyl-2-oxazolin-2-yl pyridine, s,s-ph-pybox, 2,6-bis 4s-4-phenyl-2-oxazolinyl pyridine, --2,6-bis 4s-4-phenyl-2-oxazolin-2-yl pyridine, ph-pybox, s,s, unii-5r62cxu58k, s,s-2,6-bis 4-phenyl-2-oxazolin-2-yl pyridine, pyridine, 2,6-bis 4s-4,5-dihydro-4-phenyl-2-oxazolyl, 2,6-bis 4s-4-phenyl-4,5-dihydro-1,3-oxazol-2-yl pyridine |
| PubChem CID | 7010100 |
| IUPAC Name | (4S)-4-phenyl-2-[6-[(4S)-4-phenyl-4,5-dihydro-1,3-oxazol-2-yl]pyridin-2-yl]-4,5-dihydro-1,3-oxazole |
| SMILES | C1C(N=C(O1)C2=NC(=CC=C2)C3=NC(CO3)C4=CC=CC=C4)C5=CC=CC=C5 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B2220250MG
|
TCI America
B2220250MG |
250 mg |
Each for $255.38
|
|
|||||
|
B22201G
|
TCI America
B22201G |
1 g |
Each for $743.76
|
|
|||||
Chemical Identifiers
| 174500-20-0 | |
| 369.424 | |
| HLHBIMJNCKZZQO-NHCUHLMSSA-N | |
| 7010100 | |
| C1C(N=C(O1)C2=NC(=CC=C2)C3=NC(CO3)C4=CC=CC=C4)C5=CC=CC=C5 |
| C23H19N3O2 | |
| MFCD01863586 | |
| 2,6-bis s-4-phenyl-4,5-dihydrooxazol-2-yl pyridine, 2,6-bis 4s-phenyl-2-oxazolin-2-yl pyridine, s,s-ph-pybox, 2,6-bis 4s-4-phenyl-2-oxazolinyl pyridine, --2,6-bis 4s-4-phenyl-2-oxazolin-2-yl pyridine, ph-pybox, s,s, unii-5r62cxu58k, s,s-2,6-bis 4-phenyl-2-oxazolin-2-yl pyridine, pyridine, 2,6-bis 4s-4,5-dihydro-4-phenyl-2-oxazolyl, 2,6-bis 4s-4-phenyl-4,5-dihydro-1,3-oxazol-2-yl pyridine | |
| (4S)-4-phenyl-2-[6-[(4S)-4-phenyl-4,5-dihydro-1,3-oxazol-2-yl]pyridin-2-yl]-4,5-dihydro-1,3-oxazole |
Specifications
| 174500-20-0 | |
| White-Yellow | |
| MFCD01863586 | |
| 2,6-bis s-4-phenyl-4,5-dihydrooxazol-2-yl pyridine, 2,6-bis 4s-phenyl-2-oxazolin-2-yl pyridine, s,s-ph-pybox, 2,6-bis 4s-4-phenyl-2-oxazolinyl pyridine, --2,6-bis 4s-4-phenyl-2-oxazolin-2-yl pyridine, ph-pybox, s,s, unii-5r62cxu58k, s,s-2,6-bis 4-phenyl-2-oxazolin-2-yl pyridine, pyridine, 2,6-bis 4s-4,5-dihydro-4-phenyl-2-oxazolyl, 2,6-bis 4s-4-phenyl-4,5-dihydro-1,3-oxazol-2-yl pyridine | |
| C1C(N=C(O1)C2=NC(=CC=C2)C3=NC(CO3)C4=CC=CC=C4)C5=CC=CC=C5 | |
| 369.424 | |
| 369.42 | |
| Crystalline Powder |
| 164°C | |
| C23H19N3O2 | |
| 250 mg | |
| HLHBIMJNCKZZQO-NHCUHLMSSA-N | |
| (4S)-4-phenyl-2-[6-[(4S)-4-phenyl-4,5-dihydro-1,3-oxazol-2-yl]pyridin-2-yl]-4,5-dihydro-1,3-oxazole | |
| 7010100 | |
| ≥95.0% (HPLC) | |
| (S,S)-2,6-Bis(4-phenyl-2-oxazolin-2-yl)pyridine |
Safety and Handling
TSCA : No