missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S,S)-(+)-2,5-Bis(methoxymethyl)pyrrolidine, 96%, Thermo Scientific™
Chemical Identifiers
| CAS | 93621-94-4 |
|---|---|
| Molecular Formula | C8H17NO2 |
| Molecular Weight (g/mol) | 159.23 |
| MDL Number | MFCD00210016 |
| InChI Key | HDXYTIYVVNJFLU-YUMQZZPRSA-N |
| Synonym | 2s,5s-2,5-bis methoxymethyl pyrrolidine, s,s-2,5-bis-methoxymethylpyrrolidine, s,s-2,5-bis methoxymethyl pyrrolidine, s,s-+ 2,5-bis methoxymethyl pyrrolidine, s,s-+-2,5-bis methoxymethyl pyrrolidine, 2s-2alpha,5beta-bis methoxymethyl pyrrolidine, 2s,5s---2,5-bis methoxymethyl pyrrolidine, 2s,5s-2,5-bis methoxymethyl-2,3,4,5-tetrahydropyrrole |
| PubChem CID | 7019262 |
| IUPAC Name | (2S,5S)-2,5-bis(methoxymethyl)pyrrolidine |
| SMILES | COCC1CCC(N1)COC |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC293462500
|
Thermo Scientific Chemicals
293462500 |
250mg | Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC293460010
|
Thermo Scientific Chemicals
293460010 |
1g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 93621-94-4 | |
| 159.23 | |
| HDXYTIYVVNJFLU-YUMQZZPRSA-N | |
| 7019262 | |
| COCC1CCC(N1)COC |
| C8H17NO2 | |
| MFCD00210016 | |
| 2s,5s-2,5-bis methoxymethyl pyrrolidine, s,s-2,5-bis-methoxymethylpyrrolidine, s,s-2,5-bis methoxymethyl pyrrolidine, s,s-+ 2,5-bis methoxymethyl pyrrolidine, s,s-+-2,5-bis methoxymethyl pyrrolidine, 2s-2alpha,5beta-bis methoxymethyl pyrrolidine, 2s,5s---2,5-bis methoxymethyl pyrrolidine, 2s,5s-2,5-bis methoxymethyl-2,3,4,5-tetrahydropyrrole | |
| (2S,5S)-2,5-bis(methoxymethyl)pyrrolidine |
Specifications
| 93621-94-4 | |
| 100.0 | |
| 95% min. (GC) | |
| C8H17NO2 | |
| MFCD00210016 | |
| + 8.00 | |
| HDXYTIYVVNJFLU-YUMQZZPRSA-N | |
| (2S,5S)-2,5-bis(methoxymethyl)pyrrolidine | |
| 7019262 | |
| 96% |
| 95.0 | |
| Authentic | |
| Glass Bottle | |
| 1.4470 to 1.4490 | |
| 250mg | |
| 2s,5s-2,5-bis methoxymethyl pyrrolidine, s,s-2,5-bis-methoxymethylpyrrolidine, s,s-2,5-bis methoxymethyl pyrrolidine, s,s-+ 2,5-bis methoxymethyl pyrrolidine, s,s-+-2,5-bis methoxymethyl pyrrolidine, 2s-2alpha,5beta-bis methoxymethyl pyrrolidine, 2s,5s---2,5-bis methoxymethyl pyrrolidine, 2s,5s-2,5-bis methoxymethyl-2,3,4,5-tetrahydropyrrole | |
| COCC1CCC(N1)COC | |
| 159.23 | |
| 159.23 | |
| (S,S)-(+)-2,5-Bis(methoxymethyl)pyrrolidine, 96% |
Safety and Handling
TSCA : TSCA
RUO â Research Use Only