missing translation for 'onlineSavingsMsg'
Learn More

(S,S)-(+)-2,5-Bis(methoxymethyl)pyrrolidine, 96%

Catalog No. AC293462500
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC293462500 250 mg Glass Bottle
AC293460010 1 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC293462500 Supplier Thermo Scientific Chemicals Supplier No. 293462500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 93621-94-4 | C8H17NO2 | 159.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 93621-94-4
Molecular Formula C8H17NO2
Molecular Weight (g/mol) 159.23
MDL Number MFCD00210016
InChI Key HDXYTIYVVNJFLU-YUMQZZPRSA-N
Synonym 2s,5s-2,5-bis methoxymethyl pyrrolidine, s,s-2,5-bis-methoxymethylpyrrolidine, s,s-2,5-bis methoxymethyl pyrrolidine, s,s-+ 2,5-bis methoxymethyl pyrrolidine, s,s-+-2,5-bis methoxymethyl pyrrolidine, 2s-2alpha,5beta-bis methoxymethyl pyrrolidine, 2s,5s---2,5-bis methoxymethyl pyrrolidine, 2s,5s-2,5-bis methoxymethyl-2,3,4,5-tetrahydropyrrole
PubChem CID 7019262
IUPAC Name (2S,5S)-2,5-bis(methoxymethyl)pyrrolidine
SMILES COCC1CCC(N1)COC

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4470 to 1.4490
Quantity 250 mg
Specific Rotation + 8.00
Formula Weight 159.23
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material (S,S)-(+)-2,5-Bis(methoxymethyl)pyrrolidine, 96%
Show More Show Less

RUO – Research Use Only