missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S,S)-(+)-2,3-Butanediol 97.0+%, TCI America™
$41.02 - $229.29
Chemical Identifiers
| CAS | 19132-06-0 |
|---|---|
| Molecular Formula | C4H10O2 |
| Molecular Weight (g/mol) | 90.122 |
| MDL Number | MFCD00063648 |
| InChI Key | OWBTYPJTUOEWEK-IMJSIDKUSA-N |
| Synonym | 2s,3s-butane-2,3-diol, 2s,3s-+-2,3-butanediol, s,s-2,3-butanediol, s,s-+-2,3-butanediol, s,s-butane-2,3-diol, unii-7e9uxg71s1, 2,3-butanediol, +, s,s-2,3-butylene glycol, 2,3-butanediol, s-r*,r*, 2s,3s-2,3-butanediol |
| PubChem CID | 439888 |
| ChEBI | CHEBI:16812 |
| IUPAC Name | (2S,3S)-butane-2,3-diol |
| SMILES | CC(C(C)O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B1343100MG
|
TCI America
B1343100MG |
100 mg |
Each for $41.02
|
|
|||||
|
B13431G
|
TCI America
B13431G |
1 g |
Each for $229.29
|
|
|||||
Chemical Identifiers
| 19132-06-0 | |
| 90.122 | |
| OWBTYPJTUOEWEK-IMJSIDKUSA-N | |
| 439888 | |
| (2S,3S)-butane-2,3-diol |
| C4H10O2 | |
| MFCD00063648 | |
| 2s,3s-butane-2,3-diol, 2s,3s-+-2,3-butanediol, s,s-2,3-butanediol, s,s-+-2,3-butanediol, s,s-butane-2,3-diol, unii-7e9uxg71s1, 2,3-butanediol, +, s,s-2,3-butylene glycol, 2,3-butanediol, s-r*,r*, 2s,3s-2,3-butanediol | |
| CHEBI:16812 | |
| CC(C(C)O)O |
Specifications
| 19132-06-0 | |
| 182°C | |
| MFCD00063648 | |
| 2s,3s-butane-2,3-diol, 2s,3s-+-2,3-butanediol, s,s-2,3-butanediol, s,s-+-2,3-butanediol, s,s-butane-2,3-diol, unii-7e9uxg71s1, 2,3-butanediol, +, s,s-2,3-butylene glycol, 2,3-butanediol, s-r*,r*, 2s,3s-2,3-butanediol | |
| CC(C(C)O)O | |
| 90.122 | |
| CHEBI:16812 | |
| ≥97.0% (GC) | |
| (S,S)-(+)-2,3-Butanediol |
| Yellow | |
| C4H10O2 | |
| 100 mg | |
| OWBTYPJTUOEWEK-IMJSIDKUSA-N | |
| (2S,3S)-butane-2,3-diol | |
| 439888 | |
| 90.12 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0235
TSCA : No