missing translation for 'onlineSavingsMsg'
Learn More

(S,S)-(+)-2,3-Butanediol 97.0+%, TCI America™

Catalog No. B13431G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13431G 1 g
B1343100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B13431G Supplier TCI America Supplier No. B13431G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19132-06-0
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.122
MDL Number MFCD00063648
InChI Key OWBTYPJTUOEWEK-IMJSIDKUSA-N
Synonym 2s,3s-butane-2,3-diol, 2s,3s-+-2,3-butanediol, s,s-2,3-butanediol, s,s-+-2,3-butanediol, s,s-butane-2,3-diol, unii-7e9uxg71s1, 2,3-butanediol, +, s,s-2,3-butylene glycol, 2,3-butanediol, s-r*,r*, 2s,3s-2,3-butanediol
PubChem CID 439888
ChEBI CHEBI:16812
IUPAC Name (2S,3S)-butane-2,3-diol
SMILES CC(C(C)O)O

Specifications

Color Yellow
Boiling Point 182°C
Quantity 1 g
Formula Weight 90.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material (S,S)-(+)-2,3-Butanediol
EINECSNumber (2)-0235
TSCA No