missing translation for 'onlineSavingsMsg'
Learn More

(S,S)-(-)-2,2'-Isopropylidenebis(4-tert-butyl-2-oxazoline) 97.0+%, TCI America™

Catalog No. I0567100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0567100MG 100 mg
I05671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0567100MG Supplier TCI America Supplier No. I0567100MG

Chemical Identifiers

CAS 131833-93-7
Molecular Formula C17H30N2O2
Molecular Weight (g/mol) 294.44
MDL Number MFCD00192243
InChI Key DPMGLJUMNRDNMX-UHFFFAOYNA-N
Synonym 4s,4's-2,2'-propane-2,2-diyl bis 4-tert-butyl-4,5-dihydrooxazole, s,s---2,2'-isopropylidenebis 4-tert-butyl-2-oxazoline, t-bu-box, s,s, 2,2'-isopropylidenebis 4s-4-tert-butyl-2-oxazoline, s,s---2,2-bis 4-tert-butyl-2-oxazolin-2-yl propane, s,s---2,2-dimethylmethylene bis 4-tert-butyl-2-oxazoline, 4s-4-tert-butyl-2-2-4s-4-tert-butyl-4,5-dihydro-1,3-oxazol-2-yl propan-2-yl-4,5-dihydro-1,3-oxazole, 4-tert-butyl-2-1-4-tert-butyl-4,5-dihydro-1,3-oxazol-2-yl-1-methylethyl-4,5-dihydro-1,3-oxazole, s,s-t-bu-box
PubChem CID 688208
IUPAC Name 4-tert-butyl-2-[2-(4-tert-butyl-4,5-dihydro-1,3-oxazol-2-yl)propan-2-yl]-4,5-dihydro-1,3-oxazole
SMILES CC(C)(C)C1COC(=N1)C(C)(C)C1=NC(CO1)C(C)(C)C

Specifications

Melting Point 90°C
Color White-Yellow
Quantity 100 mg
Formula Weight 294.44
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S,S)-(-)-2,2′-Isopropylidenebis(4-tert-butyl-2-oxazoline)