missing translation for 'onlineSavingsMsg'
Learn More

(S,S)-2,2'-Isopropylidenebis(4-phenyl-2-oxazoline) 95.0+%, TCI America™

Catalog No. i0582250mg
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0582250MG 250 mg
I05821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I0582250MG Supplier TCI America Supplier No. I0582250MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 131457-46-0
Molecular Formula C21H22N2O2
Molecular Weight (g/mol) 334.419
MDL Number MFCD00192245
InChI Key JTNVCJCSECAMLD-QZTJIDSGSA-N
Synonym (S,S)-2,2-Bis(4-phenyl-2-oxazolin-2-yl)propane, (S,S)-2,2′C-(Dimethylmethylene)bis(4-phenyl-2-oxazoline)
PubChem CID 676388
IUPAC Name (4S)-4-phenyl-2-[2-[(4S)-4-phenyl-4,5-dihydro-1,3-oxazol-2-yl]propan-2-yl]-4,5-dihydro-1,3-oxazole
SMILES CC(C)(C1=NC(CO1)C2=CC=CC=C2)C3=NC(CO3)C4=CC=CC=C4

Specifications

Color Yellow
Quantity 250 mg
Formula Weight 334.42
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (S,S)-2,2′-Isopropylidenebis(4-phenyl-2-oxazoline)
TSCA No
Recommended Storage Refrigerator