missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-RUCY(regR)-XylBINAP, TCI America™
$137.94 - $413.83
Chemical Identifiers
| CAS | 1312713-89-5 |
|---|---|
| Molecular Formula | C71H73ClN2O2P2Ru |
| MDL Number | MFCD20922903 |
| Synonym | RuCl[(S)-daipena][(S)-xylbinap] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
R0138200MG
|
TCI America
R0138200MG |
200 mg |
Each for $137.94
|
|
|||||
|
R01381G
|
TCI America
R01381G |
1 g |
Each for $413.83
|
|
|||||
Chemical Identifiers
| 1312713-89-5 | |
| C71H73ClN2O2P2Ru | |
| MFCD20922903 | |
| RuCl[(S)-daipena][(S)-xylbinap] |
Specifications
| 1312713-89-5 | |
| C71H73ClN2O2P2Ru | |
| 200 mg | |
| 1184.84 | |
| (S)-RUCY(regR)-XylBINAP |
| Green-Yellow | |
| MFCD20922903 | |
| RuCl[(S)-daipena][(S)-xylbinap] | |
| Crystalline Powder |
Safety and Handling
TSCA : No