missing translation for 'onlineSavingsMsg'
Learn More

(S)-Propylene Carbonate 98.0+%, TCI America™

Catalog No. p-7135154
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14865G 5 g
P148625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P14865G Supplier TCI America Supplier No. P14865G
Only null left

Chemical Identifiers

CAS 51260-39-0
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.089
MDL Number MFCD00798264
InChI Key RUOJZAUFBMNUDX-VKHMYHEASA-N
Synonym (S)-4-Methyl-1,3-dioxolan-2-one
PubChem CID 2734444
IUPAC Name (4S)-4-methyl-1,3-dioxolan-2-one
SMILES CC1COC(=O)O1

Specifications

Color Yellow
Boiling Point 105°C
Quantity 5 g
Formula Weight 102.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-Propylene Carbonate
EINECSNumber (5)-0524
RTECSNumber FF9650000
TSCA No