missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S-Propyl Thioacetate 98.0+%, TCI America™
Supplier: TCI America P171625G
Specifications
| S-Propyl Thioacetate | |
| Yellow | |
| C5H10OS | |
| 25 g | |
| s-propyl thioacetate, propyl thioacetate, ethanethioic acid, s-propyl ester, propyl thiolacetate, s-n-propyl thioacetate, acetic acid, thio-, s-propyl ester, unii-y7284nz6ep, 1-propylsulfanyl ethanone, n-propyl thio acetate, fema no. 3385 | |
| CCCSC(C)=O | |
| 118.19 | |
| 118.19 | |
| Liquid |
| 2307-10-0 | |
| 137°C | |
| MFCD00039937 | |
| 3272 | |
| SBWFWBJCYMBZEY-UHFFFAOYSA-N | |
| 1-(propylsulfanyl)ethan-1-one | |
| 61295 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 2307-10-0 | |
| 118.19 | |
| SBWFWBJCYMBZEY-UHFFFAOYSA-N | |
| 61295 | |
| CCCSC(C)=O |
| C5H10OS | |
| MFCD00039937 | |
| s-propyl thioacetate, propyl thioacetate, ethanethioic acid, s-propyl ester, propyl thiolacetate, s-n-propyl thioacetate, acetic acid, thio-, s-propyl ester, unii-y7284nz6ep, 1-propylsulfanyl ethanone, n-propyl thio acetate, fema no. 3385 | |
| 1-(propylsulfanyl)ethan-1-one |
Safety and Handling
TSCA : No