missing translation for 'onlineSavingsMsg'
Learn More

S-Propyl Thioacetate 98.0+%, TCI America™

Catalog No. P17165G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P17165G 5 g
P171625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P17165G Supplier TCI America Supplier No. P17165G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2307-10-0
Molecular Formula C5H10OS
Molecular Weight (g/mol) 118.19
MDL Number MFCD00039937
InChI Key SBWFWBJCYMBZEY-UHFFFAOYSA-N
Synonym s-propyl thioacetate, propyl thioacetate, ethanethioic acid, s-propyl ester, propyl thiolacetate, s-n-propyl thioacetate, acetic acid, thio-, s-propyl ester, unii-y7284nz6ep, 1-propylsulfanyl ethanone, n-propyl thio acetate, fema no. 3385
PubChem CID 61295
IUPAC Name 1-(propylsulfanyl)ethan-1-one
SMILES CCCSC(C)=O

Specifications

Color Yellow
Boiling Point 137°C
Quantity 5 g
UN Number 3272
Formula Weight 118.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material S-Propyl Thioacetate