missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Piperazine-2-carboxylic acid dihydrochloride, 98+%

Catalog No. AAH2897206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2897206 5 g
AAH2897203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2897206 Supplier Thermo Scientific Chemicals Supplier No. H2897206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 158663-69-5 | C5H12Cl2N2O2 | 203.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 158663-69-5
Molecular Formula C5H12Cl2N2O2
Molecular Weight (g/mol) 203.06
MDL Number MFCD01632104,MFCD03840829
InChI Key WNSDZBQLMGKPQS-UHFFFAOYNA-N
Synonym s-piperazine-2-carboxylic acid dihydrochloride, s-2-piperazinecarboxylic acid dihydrochloride, 2s-piperazine-2-carboxylic acid dihydrochloride, s-piperazine-2-carboxylic acid 2hcl, s---2-piperazinecarboxylic acid dihydrochloride, s---piperazine-2-carboxylic acid dihydrochloride, 2-piperazinecarboxylic acid, dihydrochloride, 2s, s---piperazine-2-carboxylic acid 2hcl, 2s---2-carboxypiperazine dihydrochloride
PubChem CID 2734260
IUPAC Name (2S)-piperazine-2-carboxylic acid;dihydrochloride
SMILES Cl.Cl.OC(=O)C1CNCCN1

Specifications

Melting Point ∽280°C (decomposition)
Quantity 5 g
Sensitivity Hygroscopic
Optical Rotation −5° (c=1 in Water)
Formula Weight 203.07
Percent Purity ≥98%
Chemical Name or Material (S)-(-)-Piperazine-2-carboxylic acid dihydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only