missing translation for 'onlineSavingsMsg'
Learn More

S-Phenyl Thioacetate 98.0+%, TCI America™

Catalog No. t084925ml
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
Catalog No. Quantity
T084925ML 25 mL
1 options

Catalog No. T084925ML

Supplier: TCI America T084925ML

Only null left

Chemical Identifiers

CAS 934-87-2
Molecular Formula C8H8OS
Molecular Weight (g/mol) 152.211
MDL Number MFCD00008752
InChI Key WBISVCLTLBMTDS-UHFFFAOYSA-N
Synonym s-phenyl thioacetate, thiophenyl acetate, ethanethioic acid, s-phenyl ester, acetic acid, thio-, s-phenyl ester, phenyl thioacetate, thioacetic acid s-phenyl ester, thiacetic acid, s-phenyl ester, pubchem15324, 1-phenylsulfanylethanone, thioacetic acid s-phenyl
PubChem CID 13630
IUPAC Name S-phenyl ethanethioate
SMILES CC(=O)SC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 230°C
Quantity 25 mL
Formula Weight 152.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material S-Phenyl Thioacetate
RTECSNumber AJ7559000
TSCA No