missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-Phenyl-1,2-ethanediol, 97%

CAS: 25779-13-9 | C8H10O2 | 138.166 g/mol

$58.62 - $147.93

Chemical Identifiers

CAS 25779-13-9
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00066256
InChI Key PWMWNFMRSKOCEY-MRVPVSSYSA-N
Synonym s-+-1-phenyl-1,2-ethanediol, s-1-phenylethane-1,2-diol, 1s-1-phenylethane-1,2-diol, s-+-1-phenylethane-1,2-diol, s-1-phenyl-1,2-ethanediol, unii-8ib2t1ft6b, 8ib2t1ft6b, 1,2-ethanediol, 1-phenyl-, 1s, styrene glycol, +, r---phenylethylene glycol
PubChem CID 643312
IUPAC Name (1S)-1-phenylethane-1,2-diol
SMILES C1=CC=C(C=C1)C(CO)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL09836MD
View Documents
Thermo Scientific Chemicals
L09836MD
250 mg
Each for $58.62
Only null left
 
AAL0983603
View Documents
Thermo Scientific Chemicals
L0983603
1 g
Each for $147.93
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

25779-13-9
138.166
PWMWNFMRSKOCEY-MRVPVSSYSA-N
643312
C1=CC=C(C=C1)C(CO)O
C8H10O2
MFCD00066256
s-+-1-phenyl-1,2-ethanediol, s-1-phenylethane-1,2-diol, 1s-1-phenylethane-1,2-diol, s-+-1-phenylethane-1,2-diol, s-1-phenyl-1,2-ethanediol, unii-8ib2t1ft6b, 8ib2t1ft6b, 1,2-ethanediol, 1-phenyl-, 1s, styrene glycol, +, r---phenylethylene glycol
(1S)-1-phenylethane-1,2-diol

Specifications

25779-13-9
272°C to 274°C
MFCD00066256
3196197
PWMWNFMRSKOCEY-MRVPVSSYSA-N
C1=CC=C(C=C1)C(CO)O
138.166
138.17
(S)-(+)-Phenyl-1,2-ethanediol
64°C to 66°C
C8H10O2
250 mg
s-+-1-phenyl-1,2-ethanediol, s-1-phenylethane-1,2-diol, 1s-1-phenylethane-1,2-diol, s-+-1-phenylethane-1,2-diol, s-1-phenyl-1,2-ethanediol, unii-8ib2t1ft6b, 8ib2t1ft6b, 1,2-ethanediol, 1-phenyl-, 1s, styrene glycol, +, r---phenylethylene glycol
+39° (c=3 in Ethanol)
(1S)-1-phenylethane-1,2-diol
643312
97%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only