missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-p-Toluenesulfinamide 98.0+%, TCI America™

Catalog No. T20691G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T20691G 1 g
T20695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T20691G Supplier TCI America Supplier No. T20691G

Chemical Identifiers

CAS 188447-91-8
Molecular Formula C7H9NOS
Molecular Weight (g/mol) 155.215
MDL Number MFCD06858375
InChI Key YNJDSRPIGAUCEE-JTQLQIEISA-N
Synonym s-4-methylbenzenesulfinamide, s-+-p-toluenesulfinamide, s-4-methylbezenesulfinamide, s-p-toluenesulfinamide, s-4-toluenesulfinamide, benzenesulfinamide, 4-methyl-, s, pubchem9914, s-4-methyl-benzenesulfinic acid amide, s-+-p-tolylsulfinamide, s-4-methylbenzene-1-sulfinamide
PubChem CID 11116285
IUPAC Name 4-methylbenzenesulfinamide
SMILES CC1=CC=C(C=C1)S(=O)N

Specifications

Melting Point 120°C
Quantity 1 g
Formula Weight 155.22
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(+)-p-Toluenesulfinamide
EINECSNumber (3)-1869
TSCA No