missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-p-Toluenesulfinamide, 98%

Catalog No. AC456800050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC456800050 5 g
AC456800010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC456800050 Supplier Thermo Scientific Chemicals Supplier No. 456800050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 188447-91-8 | C7H9NOS | 155.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 188447-91-8
Molecular Formula C7H9NOS
Molecular Weight (g/mol) 155.22
InChI Key YNJDSRPIGAUCEE-JTQLQIEISA-N
Synonym s-4-methylbenzenesulfinamide, s-+-p-toluenesulfinamide, s-4-methylbezenesulfinamide, s-p-toluenesulfinamide, s-4-toluenesulfinamide, benzenesulfinamide, 4-methyl-, s, pubchem9914, s-4-methyl-benzenesulfinic acid amide, s-+-p-tolylsulfinamide, s-4-methylbenzene-1-sulfinamide
PubChem CID 11116285
IUPAC Name 4-methylbenzenesulfinamide
SMILES CC1=CC=C(C=C1)S(=O)N

Specifications

Melting Point 118.0°C to 120.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Formula Weight 155.22
Percent Purity 98%
Chemical Name or Material (S)-(+)-p-Toluenesulfinamide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only