missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-NBD-APy [=(S)-(+)-4-Nitro-7-(3-aminopyrrolidin-1-yl)-2,1,3-benzoxadiazole], TCI America™

Catalog No. A5562100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5562100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A5562100MG Supplier TCI America Supplier No. A5562100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

[HPLC Labeling Reagent for e.e.Determination]

Chemical Identifiers

CAS 143112-52-1
Molecular Formula C10H11N5O3
Molecular Weight (g/mol) 249.23
MDL Number MFCD00191509
InChI Key QSDDQXROWUJAJX-UHFFFAOYNA-N
Synonym (S)-(+)-4-Nitro-7-(3-aminopyrrolidin-1-yl)benzofurazan, (S)-(+)-4-Nitro-7-(3-aminopyrrolidin-1-yl)-2,1,3-benzoxadiazole
PubChem CID 15682841
IUPAC Name 1-(7-nitro-2,1,3-benzoxadiazol-4-yl)pyrrolidin-3-amine
SMILES NC1CCN(C1)C1=CC=C(C2=NON=C12)[N+]([O-])=O

Specifications

Melting Point 175°C
Quantity 100 mg
Formula Weight 249.23
Physical Form Crystalline Powder
Chemical Name or Material (S)-(+)-NBD-APy [=(S)-(+)-4-Nitro-7-(3-aminopyrrolidin-1-yl)-2,1,3-benzoxadiazole]