missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(-)-N,N-Dimethyl-1-ferrocenylethylamine 97.0+%, TCI America™
$123.03 - $413.83
Chemical Identifiers
| CAS | 31886-57-4 |
|---|---|
| Molecular Formula | C14H19FeN |
| Molecular Weight (g/mol) | 257.16 |
| MDL Number | MFCD00066272 |
| Synonym | (S)-(-)-[1-(Dimethylamino)ethyl]ferrocene |
| IUPAC Name | (S)-(-)-N,N-Dimethyl-1-ferrocenylethylamine |
| SMILES | [Fe].c1cccc1.C[C@H](N(C)C)c1cccc1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D2529200MG
|
TCI America
D2529200MG |
200 mg |
Each for $123.03
|
|
|||||
|
D25291G
|
TCI America
D25291G |
1 g |
Each for $413.83
|
|
|||||
Chemical Identifiers
| 31886-57-4 | |
| 257.16 | |
| (S)-(-)-[1-(Dimethylamino)ethyl]ferrocene | |
| [Fe].c1cccc1.C[C@H](N(C)C)c1cccc1 |
| C14H19FeN | |
| MFCD00066272 | |
| (S)-(-)-N,N-Dimethyl-1-ferrocenylethylamine |
Specifications
| 31886-57-4 | |
| C14H19FeN | |
| 200 mg | |
| [Fe].c1cccc1.C[C@H](N(C)C)c1cccc1 | |
| 257.16 | |
| ≥97.0% (GC) | |
| (S)-(-)-N,N-Dimethyl-1-ferrocenylethylamine |
| 121°C | |
| MFCD00066272 | |
| (S)-(-)-[1-(Dimethylamino)ethyl]ferrocene | |
| (S)-(-)-N,N-Dimethyl-1-ferrocenylethylamine | |
| 257.16 | |
| Liquid |
Safety and Handling
TSCA : No