missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-N-Methyl-1-phenylethylamine 98.0+%, TCI America™

Catalog No. p-7133554
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M30471ML 1 mL
M30475ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M30471ML Supplier TCI America Supplier No. M30471ML

Chemical Identifiers

CAS 19131-99-8
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00067113
InChI Key RCSSHZGQHHEHPZ-QMMMGPOBSA-N
Synonym s---n-methyl-1-phenylethylamine, s-n-methyl-1-phenylethanamine, unii-4mu17yen7y, 4mu17yen7y, methyl 1s-1-phenylethyl amine, n-methyl-1-phenylethylamine, s, s---n,alpha-dimethylbenzylamine, n-methyl-n-1s-1-phenylethyl amine, pubchem18717
PubChem CID 2060073
IUPAC Name (1S)-N-methyl-1-phenylethanamine
SMILES CC(C1=CC=CC=C1)NC

Specifications

Boiling Point 79°C
Quantity 1 mL
UN Number 2735
Formula Weight 135.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (S)-(-)-N-Methyl-1-phenylethylamine