missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-N alpha-Dimethylbenzylamine, 97%

Catalog No. p-4812523
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC395840010 1 g Glass bottle
AC395840050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC395840010 Supplier Thermo Scientific Chemicals Supplier No. 395840010
Only null left

CAS: 19131-99-8 | C9H13N | 135.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19131-99-8
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
InChI Key RCSSHZGQHHEHPZ-QMMMGPOBSA-N
Synonym s---n-methyl-1-phenylethylamine, s-n-methyl-1-phenylethanamine, unii-4mu17yen7y, 4mu17yen7y, methyl 1s-1-phenylethyl amine, n-methyl-1-phenylethylamine, s, s---n,alpha-dimethylbenzylamine, n-methyl-n-1s-1-phenylethyl amine, pubchem18717
PubChem CID 2060073
IUPAC Name (1S)-N-methyl-1-phenylethanamine
SMILES CC(C1=CC=CC=C1)NC

Specifications

Density 0.9240g/mL
Boiling Point 74.0°C to 76.0°C (11.0 mmHg)
Flash Point 61°C
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH(CH3)NHCH3
Quantity 1 g
Specific Gravity 0.924
Solubility Information Solubility in water: insoluble
Formula Weight 135.21
Percent Purity 99+%
Chemical Name or Material (S)-(-)-N α-Dimethylbenzylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

RUO – Research Use Only