missing translation for 'onlineSavingsMsg'
Learn More

(S)-N-BOC-Piperidine-2-carboxylic acid, 98%

Catalog No. AC397830050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC397830050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC397830050 Supplier Thermo Scientific Chemicals Supplier No. 397830050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 26250-84-0 | C11H19NO4 | 229.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 26250-84-0
MDL Number MFCD00151904
InChI Key JQAOHGMPAAWWQO-QMMMGPOBSA-N
Synonym boc-pip-oh, s-1-boc-piperidine-2-carboxylic acid, s-n-boc-piperidine-2-carboxylic acid, s-1-tert-butoxycarbonyl piperidine-2-carboxylic acid, boc-l-pipecolic acid, boc-l-pip-oh, n-boc-l-pipecolinic acid, l-n-boc-pipecolic acid, 2s-1-tert-butoxycarbonyl piperidine-2-carboxylic acid, 2s-1-tert-butoxy carbonyl piperidine-2-carboxylic acid
PubChem CID 688617
IUPAC Name (2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]piperidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCCCC1C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 123.0°C to 127.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C11H19NO4
Quantity 5 g
Specific Rotation -62° to -65° (20°C, 589nm) (C=1, acetic acid)
Molecular Weight (g/mol) 229.27
Formula Weight 229.27
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (S)-N-BOC-Piperidine-2-carboxylic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only