Learn More
(S)-(-)-N-(2-Hydroxyethyl)-α-phenylethylamine, 99%, Thermo Scientific™
$103.78 - $103.78
Chemical Identifiers
| CAS | 66849-29-4 |
|---|---|
| Molecular Formula | C10H15NO |
| Molecular Weight (g/mol) | 165.23 |
| MDL Number | MFCD01862172 |
| InChI Key | GXIWMXAAPLZOBY-VIFPVBQESA-N |
| Synonym | s-2-1-phenylethyl amino ethanol, 2-1s-1-phenylethyl amino ethan-1-ol, 2-1s-1-phenylethyl amino ethanol, s---n-2-hydroxyethyl-alpha-phenylethylamine, 2-s-alpha-methylbenzyl amino ethanol, s-n-2-hydroxyethyl-a-phenylethylamine, ethanol,2-1s-1-phenylethyl amino, s-+-n-2-hydroxyethyl-phenylethylamine, s---n-2-hydroxyethyl-alpha-methylbenzylamine |
| PubChem CID | 2733847 |
| IUPAC Name | 2-[[(1S)-1-phenylethyl]amino]ethanol |
| SMILES | CC(C1=CC=CC=C1)NCCO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC338830010
|
Thermo Scientific Chemicals
338830010 |
1 g | Glass bottle |
N/A
|
|
||||
Chemical Identifiers
| 66849-29-4 | |
| 165.23 | |
| GXIWMXAAPLZOBY-VIFPVBQESA-N | |
| 2733847 | |
| CC(C1=CC=CC=C1)NCCO |
| C10H15NO | |
| MFCD01862172 | |
| s-2-1-phenylethyl amino ethanol, 2-1s-1-phenylethyl amino ethan-1-ol, 2-1s-1-phenylethyl amino ethanol, s---n-2-hydroxyethyl-alpha-phenylethylamine, 2-s-alpha-methylbenzyl amino ethanol, s-n-2-hydroxyethyl-a-phenylethylamine, ethanol,2-1s-1-phenylethyl amino, s-+-n-2-hydroxyethyl-phenylethylamine, s---n-2-hydroxyethyl-alpha-methylbenzylamine | |
| 2-[[(1S)-1-phenylethyl]amino]ethanol |
Specifications
| 66849-29-4 | |
| 100.0 | |
| Authentic | |
| Glass bottle | |
| 1.5320 to 1.5350 | |
| MFCD01862172 | |
| − 44.00 (22.00°C c=1,C2H5OH) | |
| s-2-1-phenylethyl amino ethanol, 2-1s-1-phenylethyl amino ethan-1-ol, 2-1s-1-phenylethyl amino ethanol, s---n-2-hydroxyethyl-alpha-phenylethylamine, 2-s-alpha-methylbenzyl amino ethanol, s-n-2-hydroxyethyl-a-phenylethylamine, ethanol,2-1s-1-phenylethyl amino, s-+-n-2-hydroxyethyl-phenylethylamine, s---n-2-hydroxyethyl-alpha-methylbenzylamine | |
| CC(C1=CC=CC=C1)NCCO | |
| 165.23 | |
| 165.23 | |
| Liquid |
| 98.5 | |
| Colorless | |
| 98.5% min. (GC) | |
| C10H15NO | |
| C6H5CH(CH3)NH(CH2)2OH | |
| 1 g | |
| − 44.00 | |
| GXIWMXAAPLZOBY-VIFPVBQESA-N | |
| 2-[[(1S)-1-phenylethyl]amino]ethanol | |
| 2733847 | |
| 99% | |
| (S)-(-)-N-(2-Hydroxyethyl)-α-phenylethylamine, 99% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
TSCA : TSCA
RUO – Research Use Only