missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-Mevalonic Acid Lithium Salt Analytical Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 32451-23-3 (Free Acid) |
| Molecular Formula | C6H12O4 · xLi+ |
| Linear Formula | C6H12O4 · xLi+ |
| Quantity | 10 mg |
| UN Number | NONH for all modes of transport |
| Synonym | Lithium (S)-3,5-dihydroxy-3-methyl-pentanoate; Lithium (S)-3,5-dihydroxy-3-methyl-valerate; S-MVA-Li |
| Formula Weight | 148.16 (Free Acid Basis) |
| Percent Purity | ≥97% (GC) |
| Grade | Analytical Standard |