missing translation for 'onlineSavingsMsg'
Learn More

(S)-Mevalonic Acid Lithium Salt Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022510 50 mg
111022509 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111022510 Supplier Merck Emd Millipore Supplier No. 4398750MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 32451-23-3 (Free Acid)
Molecular Formula C6H12O4 · xLi+
Linear Formula C6H12O4 · xLi+
Quantity 50 mg
UN Number NONH for all modes of transport
Synonym Lithium (S)-3,5-dihydroxy-3-methyl-pentanoate; Lithium (S)-3,5-dihydroxy-3-methyl-valerate; S-MVA-Li
Formula Weight 148.16 (Free Acid Basis)
Percent Purity ≥97% (GC)
Grade Analytical Standard