missing translation for 'onlineSavingsMsg'
Learn More

S-Methylisothiouronium sulfate, 98+%

Catalog No. p-7081184
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
250 g
500 g
2500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1104422 100 g
AAA1104430 250 g
AAA1104436 500 g
AAA110440E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 missing translation for 'options'
Catalog No. AAA1104422 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' A1104422
Only null left

CAS: 867-44-7 | C4H14N4O4S3 | 278.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 867-44-7
Molecular Formula C4H14N4O4S3
Molecular Weight (g/mol) 278.36
MDL Number MFCD00013055
InChI Key BZZXQZOBAUXLHZ-UHFFFAOYSA-N
Synonym s-methylisothiourea hemisulfate, 2-methyl-2-thiopseudourea hemisulfate, carbamimidothioic acid, methyl ester, sulfate 2:1, s-methylisothiourea sulfate 2:1, s-methylthiouronium sulfate 2:1, bis 2-methylisothiouronium sulphate, methyl carbamimidothioate sulfate 2:1, methylcarbamimidothioate sulfate 2:1, s-methylisothiourea, sulfate, 2-methyl-2-thiopseudourea sulfate 2:1
PubChem CID 13347
IUPAC Name methyl carbamimidothioate;sulfuric acid
SMILES CSC(=N)N.CSC(=N)N.OS(=O)(=O)O

Specifications

Melting Point ∼235°C (decomposition)
Density 1.28
Quantity 100 g
Beilstein 3917217
Sensitivity Hygroscopic
Formula Weight 278.38
Percent Purity ≥98%
Chemical Name or Material S-Methylisothiouronium sulfate
missing translation for 'healthHazard1' H302
missing translation for 'healthHazard2' GHS H Statement
H301
Toxic if swallowed.
missing translation for 'healthHazard3' P264b-P270-P301+P312-P330-P501c
EINECSNumber 212-759-7
RTECSNumber UM9200000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only