missing translation for 'onlineSavingsMsg'
Learn More

S-Methyl Thioacetate 95.0+%, TCI America™

Catalog No. M22865G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22865G 5 g
M228625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22865G Supplier TCI America Supplier No. M22865G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1534-08-3
Molecular Formula C3H6OS
Molecular Weight (g/mol) 90.14
MDL Number MFCD00014989
InChI Key OATSQCXMYKYFQO-UHFFFAOYSA-N
Synonym s-methyl thioacetate, methylthioacetate, ethanethioic acid, s-methyl ester, methanethiol acetate, thioacetic acid s-methyl ester, methyl thiolacetate, methyl ethanethioate, unii-pf2d4mwx79, ch3c o sch3, acetic acid, thio-, s-methyl ester
PubChem CID 73750
ChEBI CHEBI:51280
IUPAC Name S-methyl ethanethioate
SMILES CC(=O)SC

Specifications

Color Colorless
Boiling Point 98°C
Quantity 5 g
UN Number 3272
Formula Weight 90.14
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material S-Methyl Thioacetate