missing translation for 'onlineSavingsMsg'
Learn More

S-Methyl thioacetate, 98+%

Catalog No. p-7082115
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1240006 5 g
AAA1240014 25 g
AAA1240022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1240006 Supplier Thermo Scientific Chemicals Supplier No. A1240006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1534-08-3 | C3H6OS | 90.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1534-08-3
Molecular Formula C3H6OS
Molecular Weight (g/mol) 90.14
MDL Number MFCD00014989
InChI Key OATSQCXMYKYFQO-UHFFFAOYSA-N
Synonym s-methyl thioacetate, methylthioacetate, ethanethioic acid, s-methyl ester, methanethiol acetate, thioacetic acid s-methyl ester, methyl thiolacetate, methyl ethanethioate, unii-pf2d4mwx79, ch3c o sch3, acetic acid, thio-, s-methyl ester
PubChem CID 73750
ChEBI CHEBI:51280
IUPAC Name S-methyl ethanethioate
SMILES CC(=O)SC

Specifications

Density 1.024
Boiling Point 97°C to 99°C
Flash Point 12°C (54°F)
Odor Unpleasant
Quantity 5 g
UN Number UN1993
Formula Weight 91.14
Percent Purity ≥98%
Chemical Name or Material S-Methyl thioacetate
Hazard Category H225-H311+H331-H319
Hazard Statement GHS H Statement
H225-H302-H332
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P311-P312-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 216-252-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only