missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Indoline-2-carboxylic Acid 95.0+%, TCI America™

Catalog No. p-7131383
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I03951G 1 g
I03955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I03951G Supplier TCI America Supplier No. I03951G
Only null left

Chemical Identifiers

CAS 79815-20-6
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.176
MDL Number MFCD00070578
InChI Key QNRXNRGSOJZINA-QMMMGPOBSA-N
Synonym s---indoline-2-carboxylic acid, s-indoline-2-carboxylic acid, 2s-2,3-dihydro-1h-indole-2-carboxylic acid, h-idc-oh, l-indoline-2-carboxylic acid, l---indoline-2-carboxylic acid, s-2,3-dihydro-1h-indole-2-carboxylic acid, s---indolin-2-carboxylic acid, 1h-indole-2-carboxylic acid, 2,3-dihydro-, 2s
PubChem CID 2733920
IUPAC Name (2S)-2,3-dihydro-1H-indole-2-carboxylic acid
SMILES C1C(NC2=CC=CC=C21)C(=O)O

Specifications

Quantity 1 g
Formula Weight 163.18
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-Indoline-2-carboxylic Acid