missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Indoline-2-carboxylic acid, 97+%

Catalog No. p-7029537
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2700903 1 g
AAH2700906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2700903 Supplier Thermo Scientific Chemicals Supplier No. H2700903

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 79815-20-6 | C9H9NO2 | 163.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 79815-20-6
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.176
MDL Number MFCD00070578
InChI Key QNRXNRGSOJZINA-QMMMGPOBSA-N
Synonym s---indoline-2-carboxylic acid, s-indoline-2-carboxylic acid, 2s-2,3-dihydro-1h-indole-2-carboxylic acid, h-idc-oh, l-indoline-2-carboxylic acid, l---indoline-2-carboxylic acid, s-2,3-dihydro-1h-indole-2-carboxylic acid, s---indolin-2-carboxylic acid, 1h-indole-2-carboxylic acid, 2,3-dihydro-, 2s
PubChem CID 2733920
IUPAC Name (2S)-2,3-dihydro-1H-indole-2-carboxylic acid
SMILES C1C(NC2=CC=CC=C21)C(=O)O

Specifications

Melting Point ∽177°C (decomposition)
Quantity 1 g
Optical Rotation −115° (c=1 in 1N HCl)
Formula Weight 163.18
Percent Purity ≥97%
Chemical Name or Material (S)-(-)-Indoline-2-carboxylic acid
Hazard Category H317-H335-H361f-H373
Hazard Statement GHS H Statement
H361-H373-H317
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P271-P272-P280g-P281-P302+P352-P304+P340-P308+P313-P333+P313-P363-P501c
EINECSNumber 410-860-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only