Learn More
Vector Laboratories S-HyNic Linker (DMF Soluble), 5 x 1 mg
Catalog No. 502045741
Encompass
Bioconjugation. S-HyNic heterobifunctional crosslinker is fundamental to SoluLINKbioconjugation technology. S-HyNic reacts with primary amines on proteins (through lysine amino acids) or amino-modified oligonucleotides or surfaces, introducing a HyNic linker that forms stable covalent conjugates with biomolecules possessing 4FB linkers.
Catalog No. 502045741 Supplier Vector Laboratories Supplier No. S1002105
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Bioconjugation. S-HyNic heterobifunctional crosslinker is fundamental to SoluLINKbioconjugation technology. S-HyNic reacts with primary amines on proteins (through lysine amino acids) or amino-modified oligonucleotides or surfaces, introducing a HyNic linker that forms stable covalent conjugates with biomolecules possessing 4FB linkers.