missing translation for 'onlineSavingsMsg'
Learn More

(S)-Glycidyl Methyl Ether 97.0+%, TCI America™

Catalog No. p-7130225
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G02815G 5 g
G028125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G02815G Supplier TCI America Supplier No. G02815G
Only null left

Chemical Identifiers

CAS 64491-68-5
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00273366
InChI Key LKMJVFRMDSNFRT-SCSAIBSYSA-N
Synonym (S)-1-Methoxy-2,3-epoxypropane, (S)-2-(Methoxymethyl)oxirane
PubChem CID 6992187
IUPAC Name (2S)-2-(methoxymethyl)oxirane
SMILES COCC1CO1

Specifications

Color Colorless
Quantity 5 g
UN Number 1992
Formula Weight 88.11
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-Glycidyl Methyl Ether
EINECSNumber (2)-2425
RTECSNumber TZ3530000
TSCA No