missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Glycidol 96.0+%, TCI America™

Catalog No. G03645G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G03645G 5 g
G036425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G03645G Supplier TCI America Supplier No. G03645G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 60456-23-7
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.079
MDL Number MFCD00074874
InChI Key CTKINSOISVBQLD-VKHMYHEASA-N
Synonym s-glycidol, s-oxiranemethanol, s---glycidol, s-oxiran-2-ylmethanol, s---2,3-epoxy-1-propanol, 2s-oxiran-2-ylmethanol, s-+-glycidol, 2s-oxiran-2-yl methanol, oxiranemethanol, 2s
PubChem CID 6973630
ChEBI CHEBI:38690
IUPAC Name [(2S)-oxiran-2-yl]methanol
SMILES C1C(O1)CO

Specifications

Color Yellow
Boiling Point 67°C
Quantity 5 g
UN Number 2810
Formula Weight 74.08
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-(-)-Glycidol
EINECSNumber (2)-2389
RTECSNumber RR0508100
TSCA No
Recommended Storage Freezer