missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-gamma-Hydroxymethyl-gamma-butyrolactone 94.0+%, TCI America™

Catalog No. p-7130825
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09581G 1 g
H09585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H09581G Supplier TCI America Supplier No. H09581G
Only null left

Chemical Identifiers

CAS 32780-06-6
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.116
MDL Number MFCD00066224
InChI Key NSISJFFVIMQBRN-BYPYZUCNSA-N
Synonym s-5-hydroxymethyl dihydrofuran-2 3h-one, s-+-dihydro-5-hydroxymethyl-2 3h-furanone, s-+-dihydro-5-hydroxymethyl furanone, s-+-gamma-hydroxymethyl-gamma-butyrolactone, 5s-5-hydroxymethyl oxolan-2-one, s-5-hydroxymethyldihydrofuran-2-one, s-5-hydroxymethyl-dihydrofuran-2 3h-one, 2 3h-furanone, dihydro-5-hydroxymethyl-, 5s, s-4-hydroxymethyl butyrolactone, s-+-2',3'-dideoxyribonolactone
PubChem CID 2733813
IUPAC Name (5S)-5-(hydroxymethyl)oxolan-2-one
SMILES C1CC(=O)OC1CO

Specifications

Color Yellow
Quantity 1 g
Formula Weight 116.12
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-(+)-gamma-Hydroxymethyl-gamma-butyrolactone
TSCA No
Recommended Storage Freezer