missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S-Ethyl Thiopropionate 98.0+%, TCI America™
Supplier: TCI America E097125ML
Specifications
| S-Ethyl Thiopropionate | |
| -95°C | |
| 126°C | |
| MFCD00027016 | |
| 1993 | |
| HNEVHBHRLCAKKQ-UHFFFAOYSA-N | |
| S-ethyl propanethioate | |
| 75513 | |
| ≥98.0% (GC) |
| 2432-42-0 | |
| Colorless | |
| C5H10OS | |
| 25 mL | |
| s-ethyl thiopropionate, ethyl thiopropionate, propanethioic acid, s-ethyl ester, thiopropionic acid s-ethyl ester, 1-ethylsulfanyl propan-1-one, ethanethiol propionate, acmc-1ceeh, propanethioicacid,s-ethylester, thiopropionic acid, s-ethyl ester, propanethioic acid,s-ethyl ester | |
| CCC(=O)SCC | |
| 118.194 | |
| 118.19 | |
| Liquid |
Chemical Identifiers
| 2432-42-0 | |
| 118.194 | |
| HNEVHBHRLCAKKQ-UHFFFAOYSA-N | |
| 75513 | |
| CCC(=O)SCC |
| C5H10OS | |
| MFCD00027016 | |
| s-ethyl thiopropionate, ethyl thiopropionate, propanethioic acid, s-ethyl ester, thiopropionic acid s-ethyl ester, 1-ethylsulfanyl propan-1-one, ethanethiol propionate, acmc-1ceeh, propanethioicacid,s-ethylester, thiopropionic acid, s-ethyl ester, propanethioic acid,s-ethyl ester | |
| S-ethyl propanethioate |
Safety and Handling
TSCA : No