missing translation for 'onlineSavingsMsg'
Learn More

S-Ethyl Thiopropionate 98.0+%, TCI America™

Catalog No. E09715ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E09715ML 5 mL
E097125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E09715ML Supplier TCI America Supplier No. E09715ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2432-42-0
Molecular Formula C5H10OS
Molecular Weight (g/mol) 118.194
MDL Number MFCD00027016
InChI Key HNEVHBHRLCAKKQ-UHFFFAOYSA-N
Synonym s-ethyl thiopropionate, ethyl thiopropionate, propanethioic acid, s-ethyl ester, thiopropionic acid s-ethyl ester, 1-ethylsulfanyl propan-1-one, ethanethiol propionate, acmc-1ceeh, propanethioicacid,s-ethylester, thiopropionic acid, s-ethyl ester, propanethioic acid,s-ethyl ester
PubChem CID 75513
IUPAC Name S-ethyl propanethioate
SMILES CCC(=O)SCC

Specifications

Melting Point -95°C
Color Colorless
Boiling Point 126°C
Quantity 5 mL
UN Number 1993
Formula Weight 118.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material S-Ethyl Thiopropionate